Learn More
Cole-Parmer Fluke 922 Airflow Meter / Digital Micromanometer
Supplier:  Cole-Parmer 2604899
Encompass
Today’s HVAC technicians need a simple solution for diagnosing differential pressure and airflow ventilation issues. The Fluke 922 makes airflow measurements easy by combining pressure, air flow, and velocity into a single, rugged meter. Compatible with most pitot tubes, the Fluke 922 allows technicians to conveniently enter duct shape and dimensions for maximum measurement accuracy.
Catalog No. 50-630-989