Learn More
Cole-Parmer Fluke 922/KIT 922 Air Flow Meter / Digital Micromanometer
Supplier:  Cole-Parmer 0594901
Encompass
The Fluke 922 Airflow meter makes airflow measurement easy by combining three tools: differential pressure, airflow (volume) and velocity along with temperature into a single, rugged reliable meter. Great for technicians who need a simple solution for diagnosing ventilation and IAQ issues. Each parameter tells only part of the story; this meter combines all of the critical measurements in to one meter, without having to resort to expensive special application tools. You can define duct shape and size for maximum accuracy. Meter is supplied with convenient colored hoses and a piTot tube for proper pressure measurement, or use meter with most pitot tubes available in the market today. Meter also features min/max, average, hold, auto off and a bright backlit display. Available for sale in the United States and Canada only.
Catalog No. 50-630-990