Learn More
DWK Life Sciences™ Kimble™ AIRLESS-WARE™ Glass Double Oblique Manifold Plug
SDP
Catalog No. K2160509002 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Replacement Manifold Plug
Catalog No. Type
K2160509002 Replacement Manifold Plug
1 options
Catalog No. K2160509002 Supplier DWK Life Sciences™ Supplier No. 2160509002
Add to Cart
Add to Cart
Glass double oblique manifold replacement plug.
  • Replacement plug for use in Double Vacuum Gas Manifold with Metering Valves 216050-0000 and 216050-0005
  • Includes glass plug and retainer
  • Case of 1
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Manifold Plug
For Use With (Equipment) Double Vacuum Gas Manifold