Learn More
Thomson Instrument Company AirOtop Enhanced Seal for 500mL Flask
SDP
Catalog No. NC0063893
Encompass_Preferred
AirOtop Enhanced Flask Seals are sterile, hydrophobic 0.2 µm vented seals that replace loose caps and foams to deliver high oxygen transfer while maintaining a clean microbial barrier. As part of Thomson’s Ultra Yield System, customers report substantially higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•0.2 µm hydrophobic membrane for high gas exchange with reliable microbial containment
•Resealable for ~24 hours to enable sampling or additions without changing closures
•Sterile, single-use format to reduce contamination risk and cleanup
•Multiple sizes to fit common shake-flask formats
•Compatible with glass flasks and Ultra Yield Flasks
• Made in the USA
Catalog No. NC0063893 Supplier Thomson Instrument Company Supplier No. 899424
AirOtop Enhanced Flask Seals are sterile, hydrophobic 0.2 µm vented seals that replace loose caps and foams to deliver high oxygen transfer while maintaining a clean microbial barrier. As part of Thomson’s Ultra Yield System, customers report substantially higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•0.2 µm hydrophobic membrane for high gas exchange with reliable microbial containment
•Resealable for ~24 hours to enable sampling or additions without changing closures
•Sterile, single-use format to reduce contamination risk and cleanup
•Multiple sizes to fit common shake-flask formats
•Compatible with glass flasks and Ultra Yield Flasks
• Made in the USA