Learn More
Ace Glass, Inc. AirPro Condenser Reflux Coldfinger 45/50 jt for use with 5947-10 Condenser 500mm Length 45/50 jts.
SDP
Catalog No. 5000810391
Encompass_Preferred
AirPro Condenser Reflux Coldfinger 45/50 jt for use with 5947-10 Condenser 500mm Length 45/50 jts.
Catalog No. 50-008-10391 Supplier Ace Glass, Inc. Supplier No. 5947101
AirPro Condenser Reflux Coldfinger 45/50 jt for use with 5947-10 Condenser 500mm Length 45/50 jts.