Learn More
A2Z OZONE INC A2Z Ozone - A7K Air Corona Discharge Ozone Generator, 7000 mg/hour - Odor Remover
SDP
Catalog No. NC1383807
Encompass_Preferred
The A7K Air Ozone Generator stops odors from cigarette smoke and smoke damage. The A7K removes odors from skunks, pets, food, and natural smells that are hard to remove. Do not use for chemical smells such as fresh paint. The A7K includes a remote control. Remote requires 2 AAA batteries. Set 30 min, 1-hour, 2-hour, and 4-hour settings, and this unit has a 12 or 24 hour repeat timer. The unit also features single or dual plate activation for either 3500 mg or 7000 mg ozone output per hour. The unit is portable. Fan Setting Power: 95W, 98W, 100W. Air Flow Rate: 50, 100, 150 CFM. Dimensions: 10x5x11 inches. Weight: 13 lbs.
Catalog No. NC1383807 Supplier A2Z OZONE INC Supplier No. A7K
The A7K Air Ozone Generator stops odors from cigarette smoke and smoke damage. The A7K removes odors from skunks, pets, food, and natural smells that are hard to remove. Do not use for chemical smells such as fresh paint. The A7K includes a remote control. Remote requires 2 AAA batteries. Set 30 min, 1-hour, 2-hour, and 4-hour settings, and this unit has a 12 or 24 hour repeat timer. The unit also features single or dual plate activation for either 3500 mg or 7000 mg ozone output per hour. The unit is portable. Fan Setting Power: 95W, 98W, 100W. Air Flow Rate: 50, 100, 150 CFM. Dimensions: 10x5x11 inches. Weight: 13 lbs.