Learn More
Med Vet International Dynarex Disposable Guedel Airway, 80 mm, Adult Small, Green, 10/Box
Supplier:  Med Vet International 36319
Encompass_Preferred
Dynarex Resp-O2™ Guedel Airways are designed to aid the breathing process in unconscious patients by preventing the tongue from blocking the posterior pharyngeal wall. The sturdy Airways are constructed from durable, high-quality plastic materials and feature a reinforced bite block for added durability and reliability. The Airways are crafted with a flange at one end to limit the depth of insertion for optimal safety and feature smooth tapered edges to promote patient comfort and reduce mucosal trauma upon entry. The Dynarex Resp-O2™ Guedel Airway is a premium-quality respiratory device designed to help open and maintain airways in unconscious patients. The Guedel Airway’s curved construction helps to ensure an uninterrupted breathing process while minimizing patient discomfort. The Airways feature a durable bite block for added stability and are color-coded for easy size identification. 
Catalog No. 50-282-9649