Learn More
Description
- Anatomically accurate device designed for comprehensive airway management training
- Features lifelike airway anatomy, including the tongue, nasopharynx, oropharynx, epiglottis, vallecula, vocal cords, and trachea, thyroid cartilage, lungs, esophagus, and stomach for a more complete training experience
- Enables healthcare professionals, medical students and simulation centers to practice essential airway management techniques
- Helps users develop practical airway management skills for Real-world clinical situations in a controlled, safe, and effective learning environment.
- The trainer supports head tilt or chin lift and jaw thrust maneuvers
- Use of OPA, NPA, iGel, LMA, LT, ETT, NGT and Combi tube
- BVM ventilation
- Right main stem intubation
- Gastric insufflation
- Delivery Includes: Airway management trainer, Lubricant, Quick start guide, Carry bag
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Medical Training |
| Color | Dark Skin |
| Gender | Male |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


