Learn More
Med Vet International Dynarex Nasopharyngeal Airway, 36 Fr, 9.0 mm, 10/Box
Supplier:  Med Vet International 36283
Encompass_Preferred
The Dynarex Nasopharyngeal Airway is a high-quality tubular device designed to help keep patients’ airways open for improved oxygenation and ventilation. The Dynarex Nasopharyngeal Airway features a fixed flange end to limit the depth of insertion and internal striations to allow for smoother passage of suction catheters. Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes to accommodate patients’ specific needs, and packets of lubrication jelly for added convenience. Each kit comes with either 6 or 9 airways for ultimate versatility.  The Dynarex Nasopharyngeal Airway is designed for nasal insertion to help create and maintain an open and secure airway. The Dynarex Nasopharyngeal Airway features a fixed flange end for safer insertion and internal striations for optimal suctioning efficiency. Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes.
Catalog No. 50-283-1998