Learn More
Med Vet International Dynarex Oral Airways (Berman Type), 110 mm, Adult X-Large, Orange, 24/Box
Supplier:  Med Vet International 36311
Encompass_Preferred
Dynarex Resp-O2™ Oral Berman Airways are designed to aid the breathing process in unconscious patients by preventing the tongue from blocking the posterior pharyngeal wall. The Berman Airway features a dual-channel design with perforated midway openings to facilitate the insertion of suction catheters without obstructing the air passage. The Berman Airway features a smooth, curved design that contours to the shape of the pharynx for improved comfort. The Airway features a sturdy bite block for added stability and color-coding for easy size identification. The Dynarex Resp-O2™ Oral Berman Airway is a top-quality respiratory device designed to help open and maintain airways in unconscious patients. The Berman Airway is crafted with an open channel on each side and midway openings to allow access of suction catheters without obstructing patients’ airways. 
Catalog No. 50-283-1997