Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0309850
Encompass
Activation-Inducible TNF-Related Ligand (AITRL) also known as Glucocorticoid-Induced TNF-Related Ligand (GITRL) belongs to the tumor necrosis factor superfamily (TNFSF). AITRL is a Type II single transmembrane protein and shares low conservation within the extracellular domain with other TNFSF members. AITRL is expressed on macrophages immature and mature dendritic cells and B cells. Its receptor Activation-Inducible TNFR family Receptor (AITR) is expressed on T lymphocytes natural killer (NK) cells and antigen- presenting cells. After binding by AITRL AITR can be released. AITR activation increases resistance to tumors and viral infections and is involved in autoimmune and inflammatory processes. In addition activated AITR increases TCR-induced T cell proliferation and cytokine production and rescues T cells and NK cells from apoptosis.
Catalog No. 50-252-9845