Learn More
Medchemexpress LLC AITRL/TNFSF18 Human 10ug
SDP
Catalog No. 5000181161
Encompass_Preferred
AITRL a type II transmembrane protein is a ligand for glucocorticoid-induced TNFR-related protein (GITR) When AITRL binds to GITR GITR can produce costimulatory signals that regulate T-cell proliferation and effector functions GITR/AITRL interaction plays a role in the pathogenesis of tumor inflammation as well as autoimmune diseases[1] Besides AITRL plays a role in endothelial cells (EC)-activation and increases STAT-1 phosphorylation and the expression of adhesion molecules (VCAM-1 ICAM-1)[2] AITRL/TNFSF18 Protein Human is a recombinant human AITRL (E74-I198) without tag which is expressed in E coli
Catalog No. 50-001-81161 Supplier Medchemexpress LLC Supplier No. HYP731810UG
AITRL a type II transmembrane protein is a ligand for glucocorticoid-induced TNFR-related protein (GITR) When AITRL binds to GITR GITR can produce costimulatory signals that regulate T-cell proliferation and effector functions GITR/AITRL interaction plays a role in the pathogenesis of tumor inflammation as well as autoimmune diseases[1] Besides AITRL plays a role in endothelial cells (EC)-activation and increases STAT-1 phosphorylation and the expression of adhesion molecules (VCAM-1 ICAM-1)[2] AITRL/TNFSF18 Protein Human is a recombinant human AITRL (E74-I198) without tag which is expressed in E coli