Learn More
Leica Biosystems Aivia Elevate - Neuro, 3-Year Subscription
Catalog No. NC3800623
Encompass
Aivia Elevate - Neuro (3-year subscription) delivers AI-driven image analysis tools tailored for neuroscience research. The subscription provides automated 3D neuron reconstruction, dendrite and neurite segmentation, and batch-processing workflows to accelerate quantitative image analysis in research labs and core facilities.
  • AI-powered neuron reconstruction and tracing.
  • Automated 3D segmentation of neurites and dendrites.
  • Batch processing for high-throughput datasets.
  • Simplified workflows for reproducible analysis.
  • Three-year subscription term with ongoing updates and support.
Catalog No. NC3800623 Supplier Leica Biosystems Supplier No. 27100035
Add to Cart
Add to Cart
Request a Quote
Aivia Elevate - Neuro (3-year subscription) delivers AI-driven image analysis tools tailored for neuroscience research. The subscription provides automated 3D neuron reconstruction, dendrite and neurite segmentation, and batch-processing workflows to accelerate quantitative image analysis in research labs and core facilities.
  • AI-powered neuron reconstruction and tracing.
  • Automated 3D segmentation of neurites and dendrites.
  • Batch processing for high-throughput datasets.
  • Simplified workflows for reproducible analysis.
  • Three-year subscription term with ongoing updates and support.