Learn More
GSD AIX1000™ Agglutination Processor

Description
- Reads and interprets results which can be communicated via LIS
- Automated RPR Screens and Titers
- Processes 190 Samples in 90 minutes
- Complete sample traceability
- Digitally store images for retrieval anytime
- Orbital MTP Shaker features precise adherence to test procedure (100-400 RPM)
- Automatic sample location management
- Aspirates 1μl with less than 3% CV with high precision micro-syringe
- Probe-mounted high-definition camera
- Streamlined loading and decrease sample tracking errors
- Cost effective and efficient
- Multi-language support
- Proprietary digital agglutination recognition algorithm for image analysis
- LIS Compatible: RS232, USB, TCP/IP
- MS Windows Graphical User Interface (Windows 7 or later)
- 480 test kit
- 10 x 48 well plates
- 22mL Carbon Antigen
- 350μL Reactive Control
- 350μL Non-Reactive Control
Specifications
Specifications
| Type | Agglutination Processor |
| Sample Type | Serum |
| For Use With (Application) | RPR Syphilis Screening |
| For Use With (Equipment) | RPR Syphilis Kit |
| Throughput | Dispenses up to 300 μL |
| Capacity | 192 patient sample positions |
| Certifications/Compliance | FDA 510k |
| Depth (Metric) | 57 cm |
| Height (Metric) | 45/91.5 cm (Cover Closed/Cover Open) |
| Width (Metric) | 64 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.