Learn More
AJINOMOTO USA INC L-Glutamic Acid (L-Glu) 1Kg
Supplier:  AJINOMOTO USA INC 30351
Encompass
Chemical Name: (S)-2-Aminoglutaric Acid; Chemical Formula: C5H9NO4; Molecular Weight: 147.1300; Nitrogen Content: 0.09520000; CAS Number: 56-86-0; Assay: 98.5% to 100.5%; Description: White crystals or crystalline powder, slightly characteristic 1Kg
Catalog No. NC1307905