Learn More
Cayman Chemical Ajmalcn 10mg
SDP
Catalog No. 502990015
Encompass_Preferred
A terpenoid indole alkaloid; an α1-AR antagonist that reduces the phenylephrine-induced pressor response in pithed rats at 1-4 mg/kg; binds to α2A-, α2B-, α2C-, and α2D-ARs (Kis = 8.2, 14.5, 5, and 289 nM, respectively); improves hemodynamic and metabolic parameters following transient cerebral ischemia in dogs when used in combination with almitrine
Catalog No. 50-299-0015 Supplier Cayman Chemical Supplier No. 3121310 MG
A terpenoid indole alkaloid; an α1-AR antagonist that reduces the phenylephrine-induced pressor response in pithed rats at 1-4 mg/kg; binds to α2A-, α2B-, α2C-, and α2D-ARs (Kis = 8.2, 14.5, 5, and 289 nM, respectively); improves hemodynamic and metabolic parameters following transient cerebral ischemia in dogs when used in combination with almitrine