Learn More
AdipoGen Ajmalicine
SDP
Supplier:  AdipoGen CDXA0214GG25
Encompass_Preferred
Chemical. CAS 483-04-5. Formula C21H24N2O3. MW 352.43. Synthetic. Alkaloid used to study its effects as an antagonist of adrenergic and nicotinic receptors.
Catalog No. 50-256-1790