Learn More
TARGETMOL CHEMICALS INC Ajugol 20MG
SDP
Catalog No. 502876468
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Ajugol (Leonuride) shows some trypanocidal potential against Trypanosoma brucei rhodesiense (IC50 values 29.3-73.0 ug/ml). Purity 99.65%
Catalog No. 50-287-6468 Supplier TARGETMOL CHEMICALS INC Supplier No. T379220MG
Add to Cart
Add to Cart
Also available in 1 mL and bulk. Please contact Fisher for quotes. Ajugol (Leonuride) shows some trypanocidal potential against Trypanosoma brucei rhodesiense (IC50 values 29.3-73.0 ug/ml). Purity 99.65%