Learn More
Medchemexpress LLC HY-101465 5mg Medchemexpress, AK-1 CAS:330461-64-8 Purity:>98%
SDP
Catalog No. 501932142
Encompass_Preferred
Medchemexpress, HY-101465 5mg AK-1 CAS:330461-64-8 AK-1 is a potent, specific and cell-permeable SIRT2 inhibitor, with an IC 50 of 12.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2142 Supplier Medchemexpress LLC Supplier No. HY1014655MG
Medchemexpress, HY-101465 5mg AK-1 CAS:330461-64-8 AK-1 is a potent, specific and cell-permeable SIRT2 inhibitor, with an IC 50 of 12.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.