Learn More
Abcam AK-1, SIRT2 inhibitor, 10MG
Catalog No. 502705638
Encompass_Preferred
MW 403.5 Da, Purity >99%. Cell-permeable SIRT2 inhibitor (IC₅₀ = 13 μM). Increases acetylated tubulin levels. Shows neuroprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5638 Supplier Abcam Supplier No. AB14454910MG
MW 403.5 Da, Purity >99%. Cell-permeable SIRT2 inhibitor (IC₅₀ = 13 μM). Increases acetylated tubulin levels. Shows neuroprotective effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement