Learn More
Alkali Scientific AK #2 Sporulating Agar Slant 9mL Tube, 10/Pk
SDP
Catalog No. 5000662672
Encompass_Preferred
AK #2 Sporulating Agar is a specialized culture medium formulated to promote sporulation in Clostridium species and other spore forming anaerobic bacteria. Each tube contains a 9mL agar slant enriched with peptones, yeast extract, and glucose to support strong vegetative growth before encouraging spore formation as cultures mature. This balanced nutrient composition makes the medium useful for maintaining, identifying, and preserving Clostridium strains in microbiology laboratories. Compared with many standard media that support only growth, AK #2 is designed to accelerate sporulation, enabling clearer morphological identification and improved recovery of stable spore cultures. The prepared slant format provides a convenient surface for inoculation and incubation and is widely used in clinical research, food safety testing, and pharmaceutical quality control laboratories.
Catalog No. 50-006-62672 Supplier Alkali Scientific Supplier No. AKSAT00
AK #2 Sporulating Agar is a specialized culture medium formulated to promote sporulation in Clostridium species and other spore forming anaerobic bacteria. Each tube contains a 9mL agar slant enriched with peptones, yeast extract, and glucose to support strong vegetative growth before encouraging spore formation as cultures mature. This balanced nutrient composition makes the medium useful for maintaining, identifying, and preserving Clostridium strains in microbiology laboratories. Compared with many standard media that support only growth, AK #2 is designed to accelerate sporulation, enabling clearer morphological identification and improved recovery of stable spore cultures. The prepared slant format provides a convenient surface for inoculation and incubation and is widely used in clinical research, food safety testing, and pharmaceutical quality control laboratories.