Learn More
Selleck Chemical LLC AK 7
SDP
Catalog No. 5000002835
Encompass_Preferred
AK 7 is a brain-permeable selective SIRT2 inhibitor with an IC50 of 15 5 M
Catalog No. 50-000-02835 Supplier Selleck Chemical LLC Supplier No. S591410MG
AK 7 is a brain-permeable selective SIRT2 inhibitor with an IC50 of 15 5 M