Learn More
AdipoGen AK-7 (5 mg)
SDP
Supplier:  AdipoGen AGCR13511M005
Encompass_Preferred
Chemical. CAS: 420831-40-9. Formula: C19H21BrN2O3S. MW: 437.4. Brain-permeable SIRT2 (sirtuin 2) inhibitor.
Catalog No. 50-205-3889