Learn More
Medchemexpress LLC AK-778-XXMU | 1227045-76-2 | 99.9% | C22H17ClN2O3 | 1 ML
SDP
Catalog No. 5000355707
Encompass_Preferred
AK-778-XXMU is a potent DNA binding inhibitor 2 (ID2) antagonist with a KD of 129 nM. It can inhibit cell migration and invasion of glioma cell lines, induce apoptosis, and exhibits significant cancer-suppressing potency. The compound acts by inhibiting the ID2-KDR signaling axis, leading to the down-regulation of angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9), and the up-regulation of the tumor suppressor factor (PTEN). It is suitable for the study of glioma.
  • Potent ID2 antagonist with a KD of 129 nM
  • Inhibits cell migration and invasion in glioma cell lines
  • Induces apoptosis
  • Exhibits significant cancer-suppressing potency
  • Down-regulates angiogenic factors (VEGFA)
  • Down-regulates invasion-related proteins (MMP2/9)
  • Up-regulates tumor suppressor factor (PTEN)
  • Inhibits tumor growth in U87 glioma xenograft mice model
Catalog No. 50-003-55707 Supplier Medchemexpress LLC Supplier No. HY14470710MM/1ML
AK-778-XXMU is a potent DNA binding inhibitor 2 (ID2) antagonist with a KD of 129 nM. It can inhibit cell migration and invasion of glioma cell lines, induce apoptosis, and exhibits significant cancer-suppressing potency. The compound acts by inhibiting the ID2-KDR signaling axis, leading to the down-regulation of angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9), and the up-regulation of the tumor suppressor factor (PTEN). It is suitable for the study of glioma.
  • Potent ID2 antagonist with a KD of 129 nM
  • Inhibits cell migration and invasion in glioma cell lines
  • Induces apoptosis
  • Exhibits significant cancer-suppressing potency
  • Down-regulates angiogenic factors (VEGFA)
  • Down-regulates invasion-related proteins (MMP2/9)
  • Up-regulates tumor suppressor factor (PTEN)
  • Inhibits tumor growth in U87 glioma xenograft mice model