Learn More
Medchemexpress LLC AK-778-XXMU | 1227045-76-2 | 99.9% | C22H17ClN2O3 | 25 MG
SDP
Catalog No. 5000357308
Encompass_Preferred
AK-778-XXMU is a potent DNA binding inhibitor 2 (ID2) antagonist with a KD of 129 nM. It inhibits the ID2-KDR signaling axis, leading to the down-regulation of downstream angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9), while up-regulating the tumor suppressor factor (PTEN). It can inhibit cell migration and invasion of glioma cell lines, induce apoptosis, and demonstrates significant cancer-suppressing potency. This compound is used for the study of glioma.
  • Potent DNA binding inhibitor 2 (ID2) antagonist
  • Inhibits cell migration and invasion of glioma cell lines
  • Induces apoptosis
  • Demonstrates significant cancer-suppressing potency
  • Inhibits ID2-KDR signaling axis
  • Down-regulates angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9)
  • Up-regulates tumor suppressor factor (PTEN)
  • Suitable for the study of glioma
Catalog No. 50-003-57308 Supplier Medchemexpress LLC Supplier No. 25MGHY144707
AK-778-XXMU is a potent DNA binding inhibitor 2 (ID2) antagonist with a KD of 129 nM. It inhibits the ID2-KDR signaling axis, leading to the down-regulation of downstream angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9), while up-regulating the tumor suppressor factor (PTEN). It can inhibit cell migration and invasion of glioma cell lines, induce apoptosis, and demonstrates significant cancer-suppressing potency. This compound is used for the study of glioma.
  • Potent DNA binding inhibitor 2 (ID2) antagonist
  • Inhibits cell migration and invasion of glioma cell lines
  • Induces apoptosis
  • Demonstrates significant cancer-suppressing potency
  • Inhibits ID2-KDR signaling axis
  • Down-regulates angiogenic factors (VEGFA) and invasion-related proteins (MMP2/9)
  • Up-regulates tumor suppressor factor (PTEN)
  • Suitable for the study of glioma