Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ak-778-xxmu | 1227045-76-2 | MFCD16882561; MFCD08272115 | 100.0% | 392.83 g/mol | C22H17ClN2O3 | 5 MG
SDP

Catalog No. 5000221755
Encompass_Preferred

A potent small-molecule research reagent that antagonizes DNA binding protein 2 (ID2), with demonstrated binding (KD ≈129 nM) and cellular activity inhibiting glioma cell migration and inducing apoptosis.

  • Small-molecule antagonist of DNA binding protein 2 (ID2) with KD ≈129 nM.
  • Inhibits glioma cell migration and invasion, and induces apoptosis in cell models.
  • High purity (99.96%) and off-white to light yellow solid appearance.
  • Molecular formula C22H17ClN2O3; molecular weight 392.83 g/mol.
  • Recommended solvent: DMSO, 100 mg/mL (use ultrasonic and freshly opened DMSO).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution store at -80°C.

Catalog No. 50-002-21755 Supplier Medchemexpress LLC Supplier No. HY1447075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent small-molecule research reagent that antagonizes DNA binding protein 2 (ID2), with demonstrated binding (KD ≈129 nM) and cellular activity inhibiting glioma cell migration and inducing apoptosis.

  • Small-molecule antagonist of DNA binding protein 2 (ID2) with KD ≈129 nM.
  • Inhibits glioma cell migration and invasion, and induces apoptosis in cell models.
  • High purity (99.96%) and off-white to light yellow solid appearance.
  • Molecular formula C22H17ClN2O3; molecular weight 392.83 g/mol.
  • Recommended solvent: DMSO, 100 mg/mL (use ultrasonic and freshly opened DMSO).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution store at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.