Learn More
Biomatik Corporation Ak Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11033
Encompass_Preferred
Ak Polyclonal Antibody is a Penaeus monodon Polyclonal Antibody to AK. The Ak Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Penaeus monodon. Immunogen: Recombinant Penaeus Monodon Arginine Kinase Protein (2-356Aa). Applications: ELISA, WB. Target Synonyms: AK. Accession Number: C7E3T4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7623