Learn More
Enzo Life Sciences 3-O-Acetyl-11-keto-beta-boswellic acid (5mg). CAS: 67416-61-9
SDP
Supplier:  Enzo Life Sciences ALX350310M005
Encompass_Preferred
Shows antitumor and anti-inflammatory activities. Potent non-redox, noncompetitive 5-lipoxygenase and topoisomerase I and IIa inhibitor leading to apoptosis. 10-times more potent than 3-O-acetyl-β-boswellic acid (ALX-350-308). Exhibits in vivo efficacy in tumor growth and inhibition. Alternative name: AKbetaBA. Purity: ≥98% (HPLC). Solubility: Soluble in acetone, dichloromethane, diethyl ether or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8839