Learn More
McKesson General Medical AK-FLUOR Fluorescein Injection, 10% Concentration, 5 mL Volume
Catalog No. NC3595444
Encompass_Preferred
AK-FLUOR Fluorescein Injection, USP is a sterile solution designed for diagnostic fluorescein angiography and angioscopy of the retina and iris. Administered intravenously, it helps visualize ocular blood flow and identify abnormalities, ensuring precise dosing in single-dose vials. Its clear appearance and lack of preservatives make it distinctive, addressing a critical supply shortage as it transitions from Akorn Pharmaceuticals to Long Grove Pharmaceuticals.
Catalog No. NC3595444 Supplier McKesson General Medical Supplier No. 1260615
AK-FLUOR Fluorescein Injection, USP is a sterile solution designed for diagnostic fluorescein angiography and angioscopy of the retina and iris. Administered intravenously, it helps visualize ocular blood flow and identify abnormalities, ensuring precise dosing in single-dose vials. Its clear appearance and lack of preservatives make it distinctive, addressing a critical supply shortage as it transitions from Akorn Pharmaceuticals to Long Grove Pharmaceuticals.