Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK Hand-Specific Glove Dispenser

Catalog No. 17987141
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-987-141 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-987-141 Supplier AK Supplier No. AK794C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to separate left and right-handed gloves, with front access holes for easy product retrieval.

  • Separates left and right-handed gloves to minimize confusion
  • Clear design allows glove stock to be constantly monitored
  • Front access holes feature a radius edge for a smooth surface

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (L x W x H) 15 x 12 x 7.75 in.
Holds Gloves
Material PETG
No. of Compartments 2
Product Style Top Loading
Thickness (English) 0.19 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.