Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK Wipe Holder

Catalog No. 17987140E
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-987-140E White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-987-140E Supplier AK Supplier No. AK105
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Unique countertop design has slanted interior and front cut-out for fingertip access to clean wipes. Made of 3/16 in. thick (0.5cm) acrylic. Dimensions: 10 × 7 1/2 × 10 in. (25.4 × 19.1 × 25.4cm). White.

Specifications

Product Type Glove Dispenser
Color White
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Height (English) 7.5 in.
Height (Metric) 19.1 cm
Material Acrylic
For Use With (Application) Unique countertop design has slanted interior and front cut-out for fingertip access to clean wipes
Thickness (English) 0.18 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.