Learn More
Aviva Systems Biology Corp AK3 Antibody - N-terminal region (ARP56872_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56872_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AK3. Alternate names: AK3L1, AK6, AKL3L, AKL3L1, FIXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62021