Learn More
Aviva Systems Biology Corp Ak3 Antibody - C-terminal region (ARP56873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ak3. It was validated on Western Blot. . Alternate names: AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3lApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-228