Learn More
Aviva Systems Biology Corp AK3L1 Antibody - middle region (ARP41433_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41433_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AK3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AK3L1. Alternate names: AK3, AK4, AK3L1, AK3L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57851