Learn More
Aviva Systems Biology Corp AK5 Antibody - N-terminal region (ARP54829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AK5. Alternate names: AK6, MGC33326ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-54043