Learn More
ABclonal Technology AK7 Rabbit pAb
SDP
Catalog No. 5000106311
Encompass_Preferred
This gene encodes a member of the adenylate kinase family of enzymes. The encoded enzyme is a phosphotransferase that catalyzes the reversible phosphorylation of adenine nucleotides. This enzyme plays a role in energy homeostasis of the cell. Alternative splicing results in multiple transcript variants. Mutations in the mouse gene are associated with primary ciliary dyskinesia.
Catalog No. 50-001-06311 Supplier ABclonal Technology Supplier No. A14600500UL
This gene encodes a member of the adenylate kinase family of enzymes. The encoded enzyme is a phosphotransferase that catalyzes the reversible phosphorylation of adenine nucleotides. This enzyme plays a role in energy homeostasis of the cell. Alternative splicing results in multiple transcript variants. Mutations in the mouse gene are associated with primary ciliary dyskinesia.