Learn More
Medchemexpress LLC AkaLumine hydrochloride | 2558205-28-8 | 99.9% | 338.85 | 100 MG
SDP
Catalog No. 5000437586
Encompass_Preferred
AkaLumine hydrochloride is a D-luciferin analogue that emits near-infrared (NIR) light (λmax=677 nm) in reactions with native firefly luciferase (Fluc). This analogue provides high tissue-penetration and increases detection sensitivity from deep-tissue targets.
  • D-luciferin analogue
  • Emits near-infrared (NIR) light
  • High tissue-penetration
  • Increases detection sensitivity from deep-tissue targets
  • For research use only
Catalog No. 50-004-37586 Supplier Medchemexpress LLC Supplier No. HY112641A100MG
AkaLumine hydrochloride is a D-luciferin analogue that emits near-infrared (NIR) light (λmax=677 nm) in reactions with native firefly luciferase (Fluc). This analogue provides high tissue-penetration and increases detection sensitivity from deep-tissue targets.
  • D-luciferin analogue
  • Emits near-infrared (NIR) light
  • High tissue-penetration
  • Increases detection sensitivity from deep-tissue targets
  • For research use only