Learn More
Medchemexpress LLC Akalumine (hydrochloride) | 2558205-28-8 | 99.92% | 338.85 | 25 MG
SDP
Catalog No. 5000438916
Encompass_Preferred
AkaLumine hydrochloride is a D-luciferin analogue with a Km of 2.06 μM for recombinant firefly luciferase (Fluc) protein. It emits near-infrared (NIR) light (λmax=677 nm) in reactions with native Fluc, offering high tissue-penetration and increased detection sensitivity for deep-tissue targets. It is intended for research use only and not for sale to patients.
  • D-luciferin analogue
  • Km of 2.06 μM for recombinant firefly luciferase protein
  • Emits near-infrared (NIR) light (λmax=677 nm) in reactions with native Fluc
  • High tissue-penetration
  • Increases detection sensitivity from deep-tissue targets
Catalog No. 50-004-38916 Supplier Medchemexpress LLC Supplier No. 25MGHY112641A
AkaLumine hydrochloride is a D-luciferin analogue with a Km of 2.06 μM for recombinant firefly luciferase (Fluc) protein. It emits near-infrared (NIR) light (λmax=677 nm) in reactions with native Fluc, offering high tissue-penetration and increased detection sensitivity for deep-tissue targets. It is intended for research use only and not for sale to patients.
  • D-luciferin analogue
  • Km of 2.06 μM for recombinant firefly luciferase protein
  • Emits near-infrared (NIR) light (λmax=677 nm) in reactions with native Fluc
  • High tissue-penetration
  • Increases detection sensitivity from deep-tissue targets