Learn More
Aviva Systems Biology Corp AKAP1 Antibody - C-terminal region (ARP36334_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36334_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AKAP1. Alternate names: AKAP, AKAP121, AKAP149, AKAP84, D-AKAP1, MGC1807, PRKA1, SAKAP84, PPP1R43ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56118