Learn More
Aviva Systems Biology Corp AKAP10 Antibody - middle region (ARP36326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AKAP10. Alternate names: D-AKAP2, MGC9414, PRKA10ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62928