Learn More
ABclonal Technology AKAP4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148131000UL
Encompass_Preferred
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is localized to the sperm flagellum and may be involved in the regulation of sperm motility. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Catalog No. 50-001-04703