Learn More
Aviva Systems Biology Corp AKAP5 Antibody - middle region (ARP36322_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36322_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AKAP5. Alternate names: AKAP75, AKAP79, H21ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-56114