Learn More
Aviva Systems Biology Corp AKAP7 Antibody - N-terminal region (ARP36333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AKAP7. Alternate names: AKAP15, AKAP18ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-56117