Learn More
Aviva Systems Biology Corp AKAP8 Antibody - middle region (ARP32755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AKAP8. Alternate names: AKAP95, DKFZp586B1222, AKAP-8, AKAP 95, AKAP-95ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54742