Learn More
Aviva Systems Biology Corp AKAP9 Antibody - N-terminal region (ARP38677_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38677_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AKAP9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AKAP9. Alternate names: PRKA9, AKAP-9, CG-NAP, YOTIAO, AKAP350, AKAP450, PPP1R45, HYPERION, MU-RMS-40.16AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56751