Learn More
TARGETMOL CHEMICALS INC AKBA 10MG
SDP
Catalog No. 502576637
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. AKBA (3-O-Acetyl-11-keto-beta-boswellic acid) inhibits 5-lipoxygenase product formation with an IC(5) of 1.5 m muM. purity: 99%
Catalog No. 50-257-6637 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S156910MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. AKBA (3-O-Acetyl-11-keto-beta-boswellic acid) inhibits 5-lipoxygenase product formation with an IC(5) of 1.5 m muM. purity: 99%