Learn More
Medchemexpress LLC HY-N0892 5mg Medchemexpress, AKBA CAS:67416-61-9 Purity:>98%
SDP
Catalog No. 501933778
Encompass_Preferred
Medchemexpress, HY-N0892 5mg AKBA CAS:67416-61-9 AKBA (Acetyl-11-keto-β-boswellic acid) is an active triterpenoid compound from the extract of Boswellia serrate and a novel Nrf2 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3778 Supplier Medchemexpress LLC Supplier No. HYN08925MG
Medchemexpress, HY-N0892 5mg AKBA CAS:67416-61-9 AKBA (Acetyl-11-keto-β-boswellic acid) is an active triterpenoid compound from the extract of Boswellia serrate and a novel Nrf2 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.