Learn More
Medchemexpress LLC HY-123159 5mg Medchemexpress, AKI603 CAS:1432515-73-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1231595MG
Encompass_Preferred
Medchemexpress, HY-123159 5mg AKI603 CAS:1432515-73-5 AKI603 is an inhibitor of Aurora kinase A (AurA), with an IC50 of 12.3 nM. AKI603 is developed to overcome resistance mediated by BCR-ABL-T315I mutation. AKI603 exhibits strong anti-proliferative activity in leukemic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1007