Learn More
ATCC Akkermansia muciniphila
Catalog No. 502384915
Encompass
Akkermansia muciniphila strain Muc [CIP 107961] was isolated in 2002 from feces. This bacterial type strain is whole-genome sequenced and is cited to degrade mucin.
Catalog No. 50-238-4915 Supplier ATCC Supplier No. BAA835
Add to Cart
Add to Cart
Akkermansia muciniphila strain Muc [CIP 107961] was isolated in 2002 from feces. This bacterial type strain is whole-genome sequenced and is cited to degrade mucin.