Learn More
TARGETMOL CHEMICALS INC AKN-028 10MG
SDP
Catalog No. 502580103
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AKN-028 is an orally active and potent FLT3 tyrosine kinase inhibitor ( IC 50 = 6 nM). AKN-028 causes dose-dependent inhibition of FLT3 autophosphorylation. purity: 97%
Catalog No. 50-258-0103 Supplier TARGETMOL CHEMICALS INC Supplier No. T3856210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AKN-028 is an orally active and potent FLT3 tyrosine kinase inhibitor ( IC 50 = 6 nM). AKN-028 causes dose-dependent inhibition of FLT3 autophosphorylation. purity: 97%